missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 165 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 165, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6903-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003438.1 | |
Zinc finger protein 165 Protein | |
Research Use Only | |
ZNF165 | |
Recombinant Protein | |
E. coli | |
MATEPKKAAA QNSPEDEGLL IVKIEEEEFI HGQDTCLQRS ELLKQELCRQ LFRQFCYQDS PGPREALSRL RELCCQWLKP EIHTKEQILE LLVLEQFLTI LPGDLQAWVH EHYPESGEEA VTILEDLERG TDEAVLQVQA HEHGQEIFQK KVSPPGPALN VKLQPVETKA HFDSSEPQLL WDCDNESENS RSMPKLEIFE KIESQRIISG RISGYISEAS GESQDICKSA GRVKRQWEKE SGESQRLSSA QDEGFGKILT HKNTVRGEII SHDGCERRLN LNSNEFTHQK SCKHGTCDQS FKWNSDFINH QIIYAGEKNH QYGKSFKSPK LAKHAAVFSG DKTHQCNECG KAFRHSSKLA RHQRIHTGER CYECNECGKS FAESSDLTRH RRIHTGERPF GCKECGRAFN LNSHLIRHQR IHTREKPYEC SECGKTFRVS SHLIRHFRIH TGEKPYECSE CGRAFSQSSN LSQHQRIHMR ENLLM | |
Greater than 90% as determined by SDS-PAGE. |
7718 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |