missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 155 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 155, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001247415.1 |
---|---|
Gene ID (Entrez) | 7711 |
Name | Zinc finger protein 155 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16076640
|
enQuireBio™
QP6901-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096640
|
enQuireBio™
QP6901-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086640
|
enQuireBio™
QP6901-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006650
|
enQuireBio™
QP6901-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016650
|
enQuireBio™
QP6901-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026650
|
enQuireBio™
QP6901-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001247415.1 | |
Zinc finger protein 155 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7711 | |
Research Use Only | |
ZNF155 | |
Recombinant Protein | |
E. coli | |
MTTFKEAVTF KDVAVVFTEE ELGLLDPAQR KLYRDVMLEN FRNLLSVGHQ PFHQDTCHFL REEKFWMMGT ATQREGNSGG KIQTELESVP EAGAHEEWSC QQIWEQIAKD LTRSQDSIIN NSQFFENGDV PSQVEAGLPT IHTGQKPSQG GKCKQSISDV PIFDLPQQLY SEEKSYTCDE CGKSICYISA LHVHQRVHVG EKLFMCDVCG KEFSQSSHLQ THQRVHTGEK PFKCEQCGKG FSRRSALNVH RKLHTGEKPY ICEACGKAFI HDSQLKEHKR IHTGEKPFKC DICGKTFYFR SRLKSHSMVH TGEKPFRCDT CDKSFHQRSA LNRHCMVHTG EKPYRCEQCG KGFIGRLDFY KHQVVHTGEK PYNCKECGKS FRWSSCLLNH QRVHSGEKSF KCEECGKGFY TNSQLSSHQR SHSGEKPYKC EECGKGYVTK FNLDLHQRVH TGERPYNCKE CGKNFSRASS ILNHKRLHCQ KKPFKCEDCG KRLVHRTYRK DQPRDYSGEN PSKCEDCGRR YKRRLNLDIL LSLFLNDT | |
Greater than 90% as determined by SDS-PAGE. |