missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 114 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 114, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6900-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001318026.1 | |
Zinc finger protein 114 Protein | |
Research Use Only | |
ZNF114 | |
Recombinant Protein | |
E. coli | |
MSQDSVTFAD VAVNFTKEEW TLLDPAQRNL YRDVMLENSR NLAFIDWATP CKTKDATPQP DILPKRTFPE ANRVCLTSIS SQHSTLREDW RCPKTEEPHR QGVNNVKPPA VAPEKDESPV SICEDHEMRN HSKPTCRLVP SQGDSIRQCI LTRDSSIFKY NPVLNDSQKT HENNEDDGVL GWNIQWVPCG RKTELKSSTW TGSQNTVHHI RDEIDTGANR HQRNPFGKAF REDGSLRAHN THGREKMYDF TQCENTSRNN SIHAMQMQLY TAETNKKDCQ TGATSANAPN SGSHKSHCTG EKTHKCPECG RAFFYQSFLM RHMKIHTGEK PYECGKCGKA FRYSLHLNKH LRKHVVQKKP YECEECGKVI RESSKYTHIR SHTGEKPYKC KTCGKDFAKS SGLKKHLKTH KDEK | |
Greater than 90% as determined by SDS-PAGE. |
163071 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |