missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Wilm's Tumor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Wilm's Tumor 1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_000369.3 |
---|---|
Gene ID (Entrez) | 7490 |
Name | Wilm's Tumor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016580
|
enQuireBio™
QP6890-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036580
|
enQuireBio™
QP6890-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026580
|
enQuireBio™
QP6890-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046580
|
enQuireBio™
QP6890-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056580
|
enQuireBio™
QP6890-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066580
|
enQuireBio™
QP6890-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000369.3 | |
Wilm's Tumor 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7490 | |
Research Use Only | |
WT1 | |
Recombinant Protein | |
E. coli | |
MGSDVRDLNA LLPAVPSLGG GGGCALPVSG AAQWAPVLDF APPGASAYGS LGGPAPPPAP PPPPPPPPHS FIKQEPSWGG AEPHEEQCLS AFTVHFSGQF TGTAGACRYG PFGPPPPSQA SSGQARMFPN APYLPSCLES QPAIRNQGYS TVTFDGTPSY GHTPSHHAAQ FPNHSFKHED PMGQQGSLGE QQYSVPPPVY GCHTPTDSCT GSQALLLRTP YSSDNLYQMT SQLECMTWNQ MNLGATLKGV AAGSSSSVKW TEGQSNHSTG YESDNHTTPI LCGAQYRIHT HGVFRGIQDV RRVPGVAPTL VRSASETSEK RPFMCAYPGC NKRYFKLSHL QMHSRKHTGE KPYQCDFKDC ERRFSRSDQL KRHQRRHTGV KPFQCKTCQR KFSRSDHLKT HTRTHTGKTS EKPFSCRWPS CQKKFARSDE LVRHHNMHQR NMTKLQLAL | |
Greater than 90% as determined by SDS-PAGE. |