missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vitamin D-binding Protein
A DNA sequence encoding the Homo sapiens (Human) Vitamin D-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9147-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000574.2 | |
Vitamin D-binding Protein | |
Research Use Only | |
GC | |
Recombinant Protein | |
Yeast | |
RGRDYEKNKV CKEFSHLGKE DFTSLSLVLY SRKFPSGTFE QVSQLVKEVV SLTEACCAEG ADPDCYDTRT SALSAKSCES NSPFPVHPGT AECCTKEGLE RKLCMAALKH QPQEFPTYVE PTNDEICEAF RKDPKEYANQ FMWEYSTNYG QAPLSLLVSY TKSYLSMVGS CCTSASPTVC FLKERLQLKH LSLLTTLSNR VCSQYAAYGE KKSRLSNLIK LAQKVPTADL EDVLPLAEDI TNILSKCCES ASEDCMAKEL PEHTVKLCDN LSTKNSKFED CCQEKTAMDV FVCTYFMPAA QLPELPDVEL PTNKDVCDPG NTKVMDKYTF ELSRRTHLPE VFLSKVLEPT LKSLGECCDV EDSTTCFNAK GPLLKKELSS FIDKGQELCA DYSENTFTEY KKKLAERLKA KLPDATPKEL AKLVNKRSDF ASNCCSINSP PLYCDSEIDA ELKNIL | |
Greater than 90% as determined by SDS-PAGE. |
2638 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMOSTAR | |
Tris-based buffer, 50% glycerol |