missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VILIP-1 / VSNL1 Protein

A DNA sequence encoding the Homo sapiens (Human) VILIP-1 / VSNL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6883-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006540

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003376.2
VILIP-1 / VSNL1 Protein
Research Use Only
VSNL1
Recombinant Protein
E. coli
GKQNSKLAPE VMEDLVKSTE FNEHELKQWY KGFLKDCPSG RLNLEEFQQL YVKFFPYGDA SKFAQHAFRT FDKNGDGTID FREFICALSI TSRGSFEQKL NWAFNMYDLD GDGKITRVEM LEIIEAIYKM VGTVIMMKMN EDGLTPEQRV DKIFSKMDKN KDDQITLDEF KEAAKSDPSI VLLLQCDIQK
Greater than 80% as determined by SDS-PAGE.
7447
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers