missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vesicle-associated membrane protein 1
A DNA sequence encoding the Homo sapiens (Human) Vesicle-associated membrane protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6874-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_055047.2 | |
Vesicle-associated membrane protein 1 | |
Research Use Only | |
VAMP2 | |
Recombinant Protein | |
E. coli | |
MSATAATAPP AAPAGEGGPP APPPNLTSNR RLQQTQAQVD EVVDIMRVNV DKVLERDQKL SELDDRADAL QAGASQFETS AAKLKRKYWW KNLKMMIILG VICAIILIII IVYFST | |
Greater than 90% as determined by SDS-PAGE. |
6844 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |