missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vacuolar fusion protein MON1 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) Vacuolar fusion protein MON1 homolog A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7799-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001135973.1 | |
Vacuolar fusion protein MON1 homolog A Protein | |
Research Use Only | |
MON1A | |
Recombinant Protein | |
E. coli | |
DGYKVVFVRR SPLVLVAVAR TRQSAQELAQ ELLYIYYQIL SLLTGAQLSH IFQQKQNYDL RRLLSGSERI TDNLLQLMAR DPSFLMGAAR CLPLAAAVRD TVSASLQQAR ARSLVFSILL ARNQLVALVR RKDQFLHPID LHLLFNLISS SSSFREGEAW TPVCLPKFNA AGFFHAHISY LEPDTDLCLL LVSTDREDFF AVSDCRRRFQ ERLRKRGAHL ALREALRTPY YSVAQVGIPD LRHFLYKSKS SGLFTSPEIE APYTSEEEQE RLLGLYQYLH SRAHNASRPL KTIYYTGPNE NLLAWVTGAF ELYMCYSPLG TKASAVSAIH KLMRWIRKEE DRLFILTPLT Y | |
Greater than 90% as determined by SDS-PAGE. |
84315 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |