missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UV excision repair protein RAD23 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) UV excision repair protein RAD23 homolog A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6578-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257291.1 | |
UV excision repair protein RAD23 homolog A Protein | |
Research Use Only | |
RAD23A | |
Recombinant Protein | |
E. coli | |
MAVTITLKTL QQQTFKIRME PDETVKVLKE KIEAEKGRDA FPVAGQKLIY AGKILSDDVP IRDYRIDEKN FVVVMVTKTK AGQGTSAPPE ASPTAAPESS TSFPPAPTSG MSHPPPAARE DKSPSEESAP TTSPESVSGS VPSSGSSGRE EDAASTLVTG SEYETMLTEI MSMGYERERV VAALRASYNN PHRAVEYLLT GIPGSPEPEH GSVQESQVSE QPATEAAGEN PLEFLRDQPQ FQNMRQVIQQ NPALLPALLQ QLGQENPQLL QQISRHQEQF IQMLNEPPGE LADISDVEGE VGAIGEEAPQ MNYIQVTPQE KEAIERLKAL GFPESLVIQA YFACEKNENL AANFLLSQNF DDE | |
Greater than 80% as determined by SDS-PAGE. |
5886 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |