missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human USH1C Protein
A DNA sequence encoding the Homo sapiens (Human) USH1C, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001284693.1 |
---|---|
Gene ID (Entrez) | 10083 |
Name | USH1C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033771
|
enQuireBio™
QP8285-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053771
|
enQuireBio™
QP8285-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043771
|
enQuireBio™
QP8285-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063771
|
enQuireBio™
QP8285-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073771
|
enQuireBio™
QP8285-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083771
|
enQuireBio™
QP8285-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001284693.1 | |
USH1C Protein | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10083 | |
Research Use Only | |
USH1C | |
Recombinant Protein | |
E. coli | |
MDRKVAREFR HKVDFLIEND AEKDYLYDVL RMYHQTMDVA VLVGDLKLVI NEPSRLPLFD AIRPLIPLKH QVEYDQLTPR RSRKLKEVRL DRLHPEGLGL SVRGGLEFGC GLFISHLIKG GQADSVGLQV GDEIVRINGY SISSCTHEEV INLIRTKKTV SIKVRHIGLI PVKSSPDEPL TWQYVDQFVS ESGGVRGSLG SPGNRENKEK KVFISLVGSR GLGCSISSGP IQKPGIFISH VKPGSLSAEV GLEIGDQIVE VNGVDFSNLD HKEGRELFMT DRERLAEARQ RELQRQELLM QKRLAMESNK ILQEQQEMER QRRKEIAQKA AEENERYRKE MEQIVEEEEK FKKQWEEDWG SKEQLLLPKT ITAEVHPVPL RKPKYDQGVE PELEPADDLD GGTEEQGEQD FRKYEEGFDP YSMFTPEQIM GKDVRLLRIK KEGSLDLALE GGVDSPIGKV VVSAVYERGA AERHGGIVKG DEIMAINGKI VTDYTLAEAE AALQKAWNQG GDWIDLVVAV CPPKEYDDEL TFF | |
Greater than 90% as determined by SDS-PAGE. |