missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Uridine-cytidine kinase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Uridine-cytidine kinase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6867-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046410

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001129426.1
Uridine-cytidine kinase 1 Protein
Research Use Only
UCK1
Recombinant Protein
E. coli
MASAGGEDCE SPAPEADRPH QRPFLIGVSG GTASGKSTVC EKIMELLGQN EVEQRQRKVV ILSQDRFYKV LTAEQKAKAL KGQYNFDHPD AFDNDLMHRT LKNIVEGKTV EVPTYDFVTH SRLPETTVVY PADVVLFEGI LVFYSQEIRD MFHLRLFVDT DSDVRLSRRD KEVCRCDHPA RSGQYGCHQP DRAAHPGHSE W
Greater than 90% as determined by SDS-PAGE.
83549
10 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers