missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-N-acetylhexosamine pyrophosphorylase Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-N-acetylhexosamine pyrophosphorylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9752-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001311042.1 | |
UDP-N-acetylhexosamine pyrophosphorylase Protein | |
Research Use Only | |
UAP1 | |
Recombinant Protein | |
Yeast | |
MNINDLKLTL SKAGQEHLLR FWNELEEAQQ VELYAELQAM NFEELNFFFQ KAIEGFNQSS HQKNVDARME PVPREVLGSA TRDQDQLQAW ESEGLFQISQ NKVAVLLLAG GQGTRLGVAY PKGMYDVGLP SRKTLFQIQA ERILKLQQVA EKYYGNKCII PWYIMTSGRT MESTKEFFTK HKYFGLKKEN VIFFQQGMLP AMSFDGKIIL EEKNKVSMAP DGNGGLYRAL AAQNIVEDME QRGIWSIHVY CVDNILVKVA DPRFIGFCIQ KGADCGAKVV EKTNPTEPVG VVCRVDGVYQ VVEYSEISLA TAQKRSSDGR LLFNAGNIAN HFFTVPFLRD VVNVYEPQLQ HHVAQKKIPY VDTQGQLIKP DKPNGIKMEK FVFDIFQFAK KFVVYEVLRE DEFSPLKNAD SQNGKDNPTT ARHALMSLHH CWVLNAGGHF IDENGSRLPA IPRSATNGKS ETITADVNHN LKDANDVPIQ CEISPLISYA GEGLESYVAD KEFHAPLIID ENGVHELVKN GI | |
Greater than 90% as determined by SDS-PAGE. |
6675 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |