missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UDP-glucuronosyltransferase 1-4 Protein
A DNA sequence encoding the Homo sapiens (Human) UDP-glucuronosyltransferase 1-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6870-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009051.1 | |
UDP-glucuronosyltransferase 1-4 Protein | |
Research Use Only | |
UGT1A4 | |
Recombinant Protein | |
E. coli | |
GKVLVVPTDG SPWLSMREAL RELHARGHQA VVLTPEVNMH IKEEKFFTLT AYAVPWTQKE FDRVTLGYTQ GFFETEHLLK RYSRSMAIMN NVSLALHRCC VELLHNEALI RHLNATSFDV VLTDPVNLCG AVLAKYLSIP AVFFWRYIPC DLDFKGTQCP NPSSYIPKLL TTNSDHMTFL QRVKNMLYPL ALSYICHTFS APYASLASEL FQREVSVVDL VSYASVWLFR GDFVMDYPRP IMPNMVFIGG INCANGKPLS QEFEAYINAS GEHGIVVFSL GSMVSEIPEK KAMAIADALG KIPQTVLWRY TGTRPSNLAN NTILVKWLPQ NDLLGHPMTR AFITHAGSHG VYESICNGVP MVMMPLFGDQ MDNAKRMETK GAGVTLNVLE MTSEDLENAL KAVINDKSYK ENIMRLSSLH KDRPVEPLDL AVFWVEFVMR HKGAPHLRPA AHDLTWYQYH SLDVIGFLLA VVLTVAFITF KCCAYGYRKC LGKKGRVKKA HKSKTH | |
Greater than 90% as determined by SDS-PAGE. |
54657 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |