missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UCP1, mitochondrial brown fat Protein
A DNA sequence encoding the Homo sapiens (Human) UCP1, mitochondrial brown fat, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_068605.1 |
---|---|
Gene ID (Entrez) | 7350 |
Name | UCP1, mitochondrial brown fat Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16056420
|
enQuireBio™
QP6868-YE-100UG |
100 μg |
640.00 EUR
100µg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096420
|
enQuireBio™
QP6868-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066420
|
enQuireBio™
QP6868-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086420
|
enQuireBio™
QP6868-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006430
|
enQuireBio™
QP6868-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076420
|
enQuireBio™
QP6868-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068605.1 | |
UCP1, mitochondrial brown fat Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7350 | |
Research Use Only | |
UCP1 | |
Recombinant Protein | |
Yeast | |
GGLTASDVHP TLGVQLFSAG IAACLADVIT FPLDTAKVRL QVQGECPTSS VIRYKGVLGT ITAVVKTEGR MKLYSGLPAG LQRQISSASL RIGLYDTVQE FLTAGKETAP SLGSKILAGL TTGGVAVFIG QPTEVVKVRL QAQSHLHGIK PRYTGTYNAY RIIATTEGLT GLWKGTTPNL MRSVIINCTE LVTYDLMKEA FVKNNILADD VPCHLVSALI AGFCATAMSS PVDVVKTRFI NSPPGQYKSV PNCAMKVFTN EGPTAFFKGL VPSFLRLGSW NVIMFVCFEQ LKRELSKSRQ TMDCAT | |
Greater than 90% as determined by SDS-PAGE. |