missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UCHL5 Protein
A DNA sequence encoding the Homo sapiens (Human) UCHL5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6866-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186190.1 | |
UCHL5 Protein | |
Research Use Only | |
UCHL5 | |
Recombinant Protein | |
E. coli | |
MTGNAGEWCL MESDPGVFTE LIKGFGCRGA QVEEIWSLEP ENFEKLKPVH GLIFLFKWQP GEEPAGSVVQ DSRLDTIFFA KQVINNACAT QAIVSVLLNC THQDVHLGET LSEFKEFSQS FDAAMKGLAL SNSDVIRQVH NSFARQQMFE FDTKTSAKEE DAFHFVSYVP VNGRLYELDG LREGPIDLGA CNQDDWISAV RPVIEKRIQK YSEGEIRFNL MAIVSDRKMI YEQKIAELQR QLAEEPMDTD QGNSMLSAIQ SEVAKNQMLI EEEVQKLKRY KIENIRRKHN YLPFIMELLK TLAEHQQLIP LVEKFEKHFE KTLLGK | |
Greater than 90% as determined by SDS-PAGE. |
51377 | |
10 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |