missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UCHL1 / PGP9.5 Protein

A DNA sequence encoding the Homo sapiens (Human) UCHL1 / PGP9.5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8722-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026131

  • 463.00 EUR / 100µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004172.2
UCHL1 / PGP9.5 Protein
Research Use Only
UCHL1
Recombinant Protein
E. coli
MQLKPMEINP EMLNKVLSRL GVAGQWRFVD VLGLEEESLG SVPAPACALL LLFPLTAQHE NFRKKQIEEL KGQEVSPKVY FMKQTIGNSC GTIGLIHAVA NNQDKLGFED GSVLKQFLSE TEKMSPEDRA KCFEKNEAIQ AAHDAVAQEG QCRVDDKVNF HFILFNNVDG HLYELDGRMP FPVNHGASSE DTLLKDAAKV CREFTEREQG EVRFSAVALC KA
Greater than 90% as determined by SDS-PAGE.
7345
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers