missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 D3 Protein

A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 D3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6863-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026380

  • 2095.00 EUR / 1mg
Estimated Shipment: 02-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003331.1
Ubiquitin-conjugating enzyme E2 D3 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
7323
Greater than 90% as determined by SDS-PAGE.
Research Use Only
UBE2D3
Recombinant Protein
E. coli
MALKRINKEL SDLARDPPAQ CSAGPVGDDM FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TRIYHPNINS NGSICLDILR SQWSPALTIS KVLLSICSLL CDPNPDDPLV PEIARIYKTD RDKYNRISRE WTQKYAM
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers