missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tyrosinase Protein
A DNA sequence encoding the Homo sapiens (Human) Tyrosinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6860-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000363.1 | |
Tyrosinase Protein | |
Research Use Only | |
TYR | |
Recombinant Protein | |
E. coli | |
HFPRACVSSK NLMEKECCPP WSGDRSPCGQ LSGRGSCQNI LLSNAPLGPQ FPFTGVDDRE SWPSVFYNRT CQCSGNFMGF NCGNCKFGFW GPNCTERRLL VRRNIFDLSA PEKDKFFAYL TLAKHTISSD YVIPIGTYGQ MKNGSTPMFN DINIYDLFVW MHYYVSMDAL LGGSEIWRDI DFAHEAPAFL PWHRLFLLRW EQEIQKLTGD ENFTIPYWDW RDAEKCDICT DEYMGGQHPT NPNLLSPASF FSSWQIVCSR LEEYNSHQSL CNGTPEGPLR RNPGNHDKSR TPRLPSSADV EFCLSLTQYE SGSMDKAANF SFRNTLEGFA SPLTGIADAS QSSMHNALHI YMNGTMSQV | |
Greater than 90% as determined by SDS-PAGE. |
7299 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |