missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Type-1 angiotensin II receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Type-1 angiotensin II receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8960-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000676.1 | |
Type-1 angiotensin II receptor Protein | |
Research Use Only | |
AGTR1 | |
Recombinant Protein | |
Yeast | |
LNPLFYGFLG KKFKRYFLQL LKYIPPKAKS HSNLSTKMST LSYRPSDNVS SSTKKPAPCF EVE | |
Greater than 90% as determined by SDS-PAGE. |
185 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |