missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Twinkle protein, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Twinkle protein, mitochondrial, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_001157284.1 |
---|---|
Gene ID (Entrez) | 56652 |
Name | Twinkle protein, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008871
|
enQuireBio™
QP9861-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048871
|
enQuireBio™
QP9861-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018871
|
enQuireBio™
QP9861-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038871
|
enQuireBio™
QP9861-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058871
|
enQuireBio™
QP9861-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028871
|
enQuireBio™
QP9861-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001157284.1 | |
Twinkle protein, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
56652 | |
Research Use Only | |
PEO1 | |
Recombinant Protein | |
Yeast | |
APGPPRRRYR KETLQALDMP VLPVTATEIR QYLRGHGIPF QDGHSCLRAL SPFAESSQLK GQTGVTTSFS LFIDKTTGHF LCMTSLAEGS WEDFQASVEG RGDGAREGFL LSKAPEFEDS EEVRRIWNRA IPLWELPDQE EVQLADTMFG LTKVTDDTLK RFSVRYLRPA RSLVFPWFSP GGSGLRGLKL LEAKCQGDGV SYEETTIPRP SAYHNLFGLP LISRRDAEVV LTSRELDSLA LNQSTGLPTL TLPRGTTCLP PALLPYLEQF RRIVFWLGDD LRSWEAAKLF ARKLNPKRCF LVRPGDQQPR PLEALNGGFN LSRILRTALP AWHKSIVSFR QLREEVLGEL SNVEQAAGLR WSRFPDLNRI LKGHRKGELT VFTGPTGSGK TTFISEYALD LCSQGVNTLW GSFEISNVRL ARVMLTQFAE GRLEDQLDKY DHWADRFEDL PLYFMTFHGQ QSIRTVIDTM QHAVYVYDIC HVIIDNLQFM MGHEQLSTDR IAAQDYIIGV FRKFATDNNC HVTLVIHPRK EDDDKELQTA SIFGSAKASQ EADNVLILQD RKLVTGPGKR YLQVSKNRFD GDVGVFPLEF NKNSLTFSIP PKNKARLKKI KDDTGPVAKK PSSGKKGATT QNSEICSGQA PTPDQPDTSK RSK | |
Greater than 90% as determined by SDS-PAGE. |