missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tumor suppressor candidate 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Tumor suppressor candidate 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6856-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_009206.1 | |
Tumor suppressor candidate 2 Protein | |
Research Use Only | |
TUSC2 | |
Recombinant Protein | |
E. coli | |
GASGSKARGL WPFASAAGGG GSEAAGAEQA LVRPRGRAVP PFVFTRRGSM FYDEDGDLAH EFYEETIVTK NGQKRAKLRR VHKNLIPQGI VKLDHPRIHV DFPVILYEV | |
Greater than 80% as determined by SDS-PAGE. |
11334 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |