missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tumor suppressor candidate 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Tumor suppressor candidate 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6856-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046330

  • 2090.00 EUR / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_009206.1
Tumor suppressor candidate 2 Protein
Research Use Only
TUSC2
Recombinant Protein
E. coli
GASGSKARGL WPFASAAGGG GSEAAGAEQA LVRPRGRAVP PFVFTRRGSM FYDEDGDLAH EFYEETIVTK NGQKRAKLRR VHKNLIPQGI VKLDHPRIHV DFPVILYEV
Greater than 80% as determined by SDS-PAGE.
11334
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers