missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSLP Protein
A DNA sequence encoding the Homo sapiens (Human) TSLP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6845-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_149024.1 | |
TSLP Protein | |
Research Use Only | |
TSLP | |
Recombinant Protein | |
E. coli | |
YDFTNCDFEK IKAAYLSTIS KDLITYMSGT KSTEFNNTVS CSNRPHCLTE IQSLTFNPTA GCASLAKEMF AMKTKAALAI WCPGYSETQI NATQAMKKRR KRKVTTNKCL EQVSQLQGLW RRFNRPLLKQ Q | |
Greater than 90% as determined by SDS-PAGE. |
85480 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |