missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSLP Protein

A DNA sequence encoding the Homo sapiens (Human) TSLP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6845-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076240

  • 2090.00 EUR / 1mg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_149024.1
TSLP Protein
Research Use Only
TSLP
Recombinant Protein
E. coli
YDFTNCDFEK IKAAYLSTIS KDLITYMSGT KSTEFNNTVS CSNRPHCLTE IQSLTFNPTA GCASLAKEMF AMKTKAALAI WCPGYSETQI NATQAMKKRR KRKVTTNKCL EQVSQLQGLW RRFNRPLLKQ Q
Greater than 90% as determined by SDS-PAGE.
85480
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers