missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tryptase beta-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Tryptase beta-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6832-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003285.2 | |
Tryptase beta-2 Protein | |
Research Use Only | |
TPSB2 | |
Recombinant Protein | |
E. coli | |
IVGGQEAPRS KWPWQVSLRV HGPYWMHFCG GSLIHPQWVL TAAHCVGPDV KDLAALRVQL REQHLYYQDQ LLPVSRIIVH PQFYTAQIGA DIALLELEEP VNVSSHVHTV TLPPASETFP PGMPCWVTGW GDVDNDERLP PPFPLKQVKV PIMENHICDA KYHLGAYTGD DVRIVRDDML CAGNTRRDSC QGDSGGPLVC KVNGTWLQAG VVSWGEGCAQ PNRPGIYTRV TYYLDWIHHY VPKKP | |
Greater than 90% as determined by SDS-PAGE. |
64499 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |