missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Troponin C, skeletal muscle Protein
A DNA sequence encoding the Homo sapiens (Human) Troponin C, skeletal muscle, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6820-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003270.1 | |
Troponin C, skeletal muscle Protein | |
Research Use Only | |
TNNC2 | |
Recombinant Protein | |
E. coli | |
TDQQAEARSY LSEEMIAEFK AAFDMFDADG GGDISVKELG TVMRMLGQTP TKEELDAIIE EVDEDGSGTI DFEEFLVMMV RQMKEDAKGK SEEELAECFR IFDRNADGYI DPEELAEIFR ASGEHVTDEE IESLMKDGDK NNDGRIDFDE FLKMMEGVQ | |
Greater than 80% as determined by SDS-PAGE. |
7125 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |