missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Troponin C, skeletal muscle Protein

A DNA sequence encoding the Homo sapiens (Human) Troponin C, skeletal muscle, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6820-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016070

  • 463.00 EUR / 100µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003270.1
Troponin C, skeletal muscle Protein
Research Use Only
TNNC2
Recombinant Protein
E. coli
TDQQAEARSY LSEEMIAEFK AAFDMFDADG GGDISVKELG TVMRMLGQTP TKEELDAIIE EVDEDGSGTI DFEEFLVMMV RQMKEDAKGK SEEELAECFR IFDRNADGYI DPEELAEIFR ASGEHVTDEE IESLMKDGDK NNDGRIDFDE FLKMMEGVQ
Greater than 80% as determined by SDS-PAGE.
7125
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers