missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Trimethylguanosine synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Trimethylguanosine synthase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7949-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031791

  • 463.00 EUR / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001304831.1
Trimethylguanosine synthase Protein
Research Use Only
TGS1
Recombinant Protein
E. coli
MRVIAIDIDP VKIALARNNA EVYGIADKIE FICGDFLLLA SFLKADVVFL SPPWGGPDYA TAETFDIRTM MSPDGFEIFR LSKKITNNIV YFLPRNADID QVASLAGPGG QVEIEQNFLN NKLKTITAYF GDLIRRPASE T
Greater than 90% as determined by SDS-PAGE.
96764
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers