missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TREM2 Protein
A DNA sequence encoding the Homo sapiens (Human) TREM2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6839-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001258750.1 | |
TREM2 Protein | |
Research Use Only | |
Trem2 | |
Recombinant Protein | |
Yeast | |
HNTTVFQGVA GQSLQVSCPY DSMKHWGRRK AWCRQLGEKG PCQRVVSTHN LWLLSFLRRW NGSTAITDDT LGGTLTITLR NLQPHDAGLY QCQSLHGSEA DTLRKVLVEV LADPLDHRDA GDLWFPGESE SFEDAHVEHS ISRSLLEGEI PFPPTS | |
Greater than 90% as determined by SDS-PAGE. |
54209 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |