missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane protein 98 Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane protein 98, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6807-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001028676.1 | |
Transmembrane protein 98 Protein | |
Research Use Only | |
TMEM98 | |
Recombinant Protein | |
E. coli | |
LVCRQRYCRP RDLLQRYDSK PIVDLIGAME TQSEPSELEL DDVVITNPHI EAILENEDWI EDASGLMSHC IAILKICHTL TEKLVAMTMG SGAKMKTSAS VSDIIVVAKR ISPRVDDVVK SMYPPLDPKL LDARTTALLL SVSHLVLVTR NACHLTGGLD WIDQSLSAAE EHLEVLREAA LASEPDKGLP GPEGFLQEQS AI | |
Greater than 90% as determined by SDS-PAGE. |
26022 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |