missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane 4 L6 family member 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane 4 L6 family member 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9438-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055035.1 | |
Transmembrane 4 L6 family member 1 Protein | |
Research Use Only | |
TM4SF1 | |
Recombinant Protein | |
Yeast | |
LAEGPLCLDS LGQWNYTFAS TEGQYLLDTS TWSECTEPKH IVEWNVS | |
Greater than 90% as determined by SDS-PAGE. |
4071 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |