missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional regulator ERG Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional regulator ERG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9126-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001129626.1 | |
Transcriptional regulator ERG Protein | |
Research Use Only | |
ERG | |
Recombinant Protein | |
Yeast | |
MIQTVPDPAA HIKEALSVVS EDQSLFECAY GTPHLAKTEM TASSSSDYGQ TSKMSPRVPQ QDWLSQPPAR VTIKMECNPS QVNGSRNSPD ECSVAKGGKM VGSPDTVGMN YGSYMEEKHM PPPNMTTNER RVIVPADPTL WSTDHVRQWL EWAVKEYGLP DVNILLFQNI DGKELCKMTK DDFQRLTPSY NADILLSHLH YLRETPLPHL TSDDVDKALQ NSPRLMHARN TGGAAFIFPN TSVYPEATQR ITTRPDLPYE PPRRSAWTGH GHPTPQSKAA QPSPSTVPKT EDQRPQLDPY QILGPTSSRL ANPGSGQIQL WQFLLELLSD SSNSSCITWE GTNGEFKMTD PDEVARRWGE RKSKPNMNYD KLSRALRYYY DKNIMTKVHG KRYAYKFDFH GIAQALQPHP PESSLYKYPS DLPYMGSYHA HPQKMNFVAP HPPALPVTSS SFFAAPNPYW NSPTGGIYPN TRLPTSHMPS HLGTYY | |
Greater than 90% as determined by SDS-PAGE. |
2078 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |