missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional adapter 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcriptional adapter 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8449-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024681

  • 458.00 EUR / 100µg
Estimated Shipment: 27-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265199.1
Transcriptional adapter 3 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
10474
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TADA3
Recombinant Protein
E. coli
MSELKDCPLQ FHDFKSVDHL KVCPRYTAVL ARSEDDGIGI EELDTLQLEL ETLLSSASRR LRVLEAETQI LTDWQDKKGD RRFLKLGRDH ELGAPPKHGK PKKQKLEGKA GHGPGPGPGR PKSKNLQPKI QEYEFTDDPI DVPRIPKNDA PNRFWASVEP YCADITSEEV RTLEELLKPP EDEAEHYKIP PLGKHYSQRW AQEDLLEEQK DGARAAAVAD KKKGLMGPLT ELDTKDVD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers