missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcription factor BTF3 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcription factor BTF3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8384-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024341

  • 2090.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001032726.1
Transcription factor BTF3 Protein
Research Use Only
BTF3
Recombinant Protein
E. coli
TIMNQEKLAK LQAQVRIGGK GTARRKKKVV HRTATADDKK LQFSLKKLGV NNISGIEEVN MFTNQGTVIH FNNPKVQASL AANTFTITGH AETKQLTEML PSILNQLGAD SLTSLRRLAE ALPKQSVDGK APLATGEDDD DEVPDLVENF DEASKNEAN
Greater than 90% as determined by SDS-PAGE.
689
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers