missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TP53-regulated inhibitor of apoptosis 1 Protein
A DNA sequence encoding the Homo sapiens (Human) TP53-regulated inhibitor of apoptosis 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6841-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057483.1 | |
TP53-regulated inhibitor of apoptosis 1 Protein | |
Research Use Only | |
TRIAP1 | |
Recombinant Protein | |
E. coli | |
MNSVGEACTD MKREYDQCFN RWFAEKFLKG DSSGDPCTDL FKRYQQCVQK AIKEKEIPIE GLEFMGHGKE KPENSS | |
Greater than 80% as determined by SDS-PAGE. |
51499 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |