missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TP53-regulated inhibitor of apoptosis 1 Protein

A DNA sequence encoding the Homo sapiens (Human) TP53-regulated inhibitor of apoptosis 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6841-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076210

  • 2090.00 EUR / 1mg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057483.1
TP53-regulated inhibitor of apoptosis 1 Protein
Research Use Only
TRIAP1
Recombinant Protein
E. coli
MNSVGEACTD MKREYDQCFN RWFAEKFLKG DSSGDPCTDL FKRYQQCVQK AIKEKEIPIE GLEFMGHGKE KPENSS
Greater than 80% as determined by SDS-PAGE.
51499
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers