missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFSF10 / TRAIL / APO-2L Protein

A DNA sequence encoding the Homo sapiens (Human) TNFSF10 / TRAIL / APO-2L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6817-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16086050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001177871.1
TNFSF10 / TRAIL / APO-2L Protein
Research Use Only
TNFSF10
Recombinant Protein
E. coli
TNELKQMQDK YSKSGIACFL KEDDSYWDPN DEESMNSPCW QVKWQLRQLV RKMILRTSEE TISTVQEKQQ NISPLVRERG PQRVAAHITG TRGRSNTLSS PNSKNEKALG RKINSWESSR SGHSFLSNLH LRNGELVIHE KGFYYIYSQT YFRFQEEIKE NTKNDKQMVQ YIYKYTSYPD PILLMKSARN SCWSKDAEYG LYSIYQGGIF ELKENDRIFV SVTNEHLIDM DHEASFFGAF LVG
Greater than 90% as determined by SDS-PAGE.
8743
50 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers