missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFAIP6 Protein

A DNA sequence encoding the Homo sapiens (Human) TNFAIP6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6813-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_009046.2
TNFAIP6 Protein
Research Use Only
TNFAIP6
Recombinant Protein
E. coli
WGFKDGIFHN SIWLERAAGV YHREARSGKY KLTYAEAKAV CEFEGGHLAT YKQLEAARKI GFHVCAAGWM AKGRVGYPIV KPGPNCGFGK TGIIDYGIRL NRSERWDAYC YNPHAKECGG VFTDPKQIFK SPGFPNEYED NQICYWHIRL KYGQRIHLSF LDFDLEDDPG CLADYVEIYD SYDDVHGFVG RYCGDELPDD IISTGNVMTL KFLSDASVTA GGFQIKYVAM DPVSKSSQGK NTSTTSTGNK NFLAGRFSHL
Greater than 90% as determined by SDS-PAGE.
7130
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales