missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TMEM25 Protein
A DNA sequence encoding the Homo sapiens (Human) TMEM25, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6805-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137506.1 | |
TMEM25 Protein | |
Research Use Only | |
TMEM25 | |
Recombinant Protein | |
E. coli | |
ELEPQIDGQT WAERALRENE RHAFTCRVAG GPGTPRLAWY LDGQLQEAST SRLLSVGGEA FSGGTSTFTV TAHRAQHELN CSLQDPRSGR SANASVILNV QFKPEIAQVG AKYQEAQGPG LLVVLFALVR ANPPANVTWI DQDGPVTVNT SDFLVLDAQN YPWLTNHTVQ LQLRSLAHNL SVVATNDVGV TSASLPAPGP SRHPSLISSD SNNLKLNNVR LPRENMSLPS NLQLNDLTPD SRAVKPADRQ MAQNNSRPEL LDPEPGGLLT SQGFIRLPVL GYIYRVSSVS SDEIWL | |
Greater than 90% as determined by SDS-PAGE. |
84866 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |