missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TLR2 / CD282 Protein
A DNA sequence encoding the Homo sapiens (Human) TLR2 / CD282, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001305716.1 |
---|---|
Gene ID (Entrez) | 7097 |
Name | TLR2 / CD282 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055940
|
enQuireBio™
QP6800-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075940
|
enQuireBio™
QP6800-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16065940
|
enQuireBio™
QP6800-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085940
|
enQuireBio™
QP6800-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095940
|
enQuireBio™
QP6800-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005950
|
enQuireBio™
QP6800-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001305716.1 | |
TLR2 / CD282 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7097 | |
Research Use Only | |
TLR2 | |
Recombinant Protein | |
E. coli | |
KEESSNQASL SCDRNGICKG SSGSLNSIPS GLTEAVKSLD LSNNRITYIS NSDLQRCVNL QALVLTSNGI NTIEEDSFSS LGSLEHLDLS YNYLSNLSSS WFKPLSSLTF LNLLGNPYKT LGETSLFSHL TKLQILRVGN MDTFTKIQRK DFAGLTFLEE LEIDASDLQS YEPKSLKSIQ NVSHLILHMK QHILLLEIFV DVTSSVECLE LRDTDLDTFH FSELSTGETN SLIKKFTFRN VKITDESLFQ VMKLLNQISG LLELEFDDCT LNGVGNFRAS DNDRVIDPGK VETLTIRRLH IPRFYLFYDL STLYSLTERV KRITVENSKV FLVPCLLSQH LKSLEYLDLS ENLMVEEYLK NSACEDAWPS LQTLILRQNH LASLEKTGET LLTLKNLTNI DISKNSFHSM PETCQWPEKM KYLNLSSTRI HSVTGCIPKT LEILDVSNNN LNLFSLNLPQ LKELYISRNK LMTLPDASLL PMLLVLKISR NAITTFSKEQ LDSFHTLKTL EAGGNNFICS CEFLSFTQEQ QALAKVLIDW PANYLCDSPS HVRGQQVQDV RLSVSECHRT | |
Greater than 90% as determined by SDS-PAGE. |