missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIRAP Protein
A DNA sequence encoding the Homo sapiens (Human) TIRAP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6797-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034750.1 | |
TIRAP Protein | |
Research Use Only | |
TIRAP | |
Recombinant Protein | |
E. coli | |
MASSTSLPAP GSRPKKPLGK MADWFRQTLL KKPKKRPNSP ESTSSDASQP TSQDSPLPPS LSSVTSPSLP PTHASDSGSS RWSKDYDVCV CHSEEDLVAA QDLVSYLEGS TASLRCFLQL RDATPGGAIV SELCQALSSS HCRVLLITPG FLQDPWCKYQ MLQALTEAPG AEGCTIPLLS GLSRAAYPPE LRFMYYVDGR GPDGGFRQVK EAVMRYLQTL S | |
Greater than 80% as determined by SDS-PAGE. |
114609 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |