missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMP-3 Protein
A DNA sequence encoding the Homo sapiens (Human) TIMP-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8728-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000353.1 | |
TIMP-3 Protein | |
Research Use Only | |
TIMP3 | |
Recombinant Protein | |
E. coli | |
HPQDAFCNSD IVIRAKVVGK KLVKEGPFGT LVYTIKQMKM YRGFTKMPHV QYIHTEASES LCGLKLEVNK YQYLLTGRVY DGKMYTGLCN FVERWDQLTL SQRKGLNYRY HLGCNCKIKS CYYLPCFVTS KNECLWTDML SNFGYPGYQS KHYACIRQKG GYCSWYRGWA PPDKSIINA | |
Greater than 80% as determined by SDS-PAGE. |
7078 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |