missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIGIT / VSTM3 Protein
A DNA sequence encoding the Homo sapiens (Human) TIGIT / VSTM3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7657-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_776160.2 | |
TIGIT / VSTM3 Protein | |
Research Use Only | |
TIGIT | |
Recombinant Protein | |
E. coli | |
MMTGTIETTG NISAEKGGSI ILQCHLSSTT AQVTQVNWEQ QDQLLAICNA DLGWHISPSF KDRVAPGPGL GLTLQSLTVN DTGEYFCIYH TYPDGTYTGR IFLEVLESSV AEHGARFQIP | |
Greater than 90% as determined by SDS-PAGE. |
201633 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |