missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thyrotropin subunit beta Protein

A DNA sequence encoding the Homo sapiens (Human) Thyrotropin subunit beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6844-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006240

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000540.2
Thyrotropin subunit beta Protein
Research Use Only
TSHB
Recombinant Protein
E. coli
FCIPTEYTMH IERRECAYCL TINTTICAGY CMTRDINGKL FLPKYALSQD VCTYRDFIYR TVEIPGCPLH VAPYFSYPVA LSCKCGKCNT DYSDCIHEAI KTNYCTKPQK SY
Greater than 90% as determined by SDS-PAGE.
7252
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers