missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thioredoxin-related transmembrane protein 1

A DNA sequence encoding the Homo sapiens (Human) Thioredoxin-related transmembrane protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6810-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006010

  • 234.00 EUR / 50µg


Estimated Shipment
17-02-2020
Add to basket

Specifications

Specifications

NP_001137484.1
Thioredoxin-related transmembrane protein 1
50 μg
Not determined.
Extracellular Domain
Human
GST
Tris-based buffer, 50% glycerol
51075
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TMX2
Recombinant Protein
E. coli
LPTQREDGNP CDFDWREVEI LMFLSAIVMM KNRRSITVEQ HIGNIFMFSK VANT
Videos
Documents