Learn More
enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 12 Protein
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin domain-containing protein 12, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6857-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16006340
  • 2090.00 EUR / 1mg
Estimated Shipment: 03-07-2024
Specifications
NP_056997.1
Thioredoxin domain-containing protein 12 Protein
Research Use Only
TXNDC12
Recombinant Protein
E. coli
HNGLGKGFGD HIHWRTLEDG KKEAAASGLP LMVIIHKSWC GACKALKPKF AESTEISELS HNFVMVNLED EEEPKDEDFS PDGGYIPRIL FLDPSGKVHP EIINENGNPS YKYFYVSAEQ VVQGMKEAQE RLTGDAFRKK HLEDEL
Greater than 90% as determined by SDS-PAGE.
51060
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol