missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human THAP domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) THAP domain-containing protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7886-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001304720.1
THAP domain-containing protein 1
Research Use Only
THAP6
Recombinant Protein
E. coli
MVKCCSAIGC ASRCLPNSKL KGLTFHVFPT DENIKRKWVL AMKRLDVNAA GIWEPKKGDV LCSRHFKKTD FDRSAPNIKL KPGVIPSIFD SPYHLQGKRE KLHCRKNFTL KTVPATNYNH HLVGASSCIE EFQSQFIFEH SYSVMDSPKK LKHKLDHVIG ELEDTKESLR NVLDREKRFQ KSLRKTIREL KDECLISQET ANRLDTFCWD CCQESIEQDY IS
Greater than 90% as determined by SDS-PAGE.
152815
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali