missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFB1-induced anti-apoptotic factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) TGFB1-induced anti-apoptotic factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6790-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_004731.2
TGFB1-induced anti-apoptotic factor 1 Protein
Research Use Only
TIAF1
Recombinant Protein
E. coli
MSSPSSPFRE QSFLCAAGDA GEESRVQVLK NEVRRGSPVL LGWVEQAYAD KCVCGPSAPP APTPPSLSQR VMCNDLFKVN PFQLQQFRAD PSTASLLLCP GGLDHKLNLR GKAWG
Greater than 80% as determined by SDS-PAGE.
9220
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers