missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFB1-induced anti-apoptotic factor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) TGFB1-induced anti-apoptotic factor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6790-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015880

  • 2090.00 EUR / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004731.2
TGFB1-induced anti-apoptotic factor 1 Protein
Research Use Only
TIAF1
Recombinant Protein
E. coli
MSSPSSPFRE QSFLCAAGDA GEESRVQVLK NEVRRGSPVL LGWVEQAYAD KCVCGPSAPP APTPPSLSQR VMCNDLFKVN PFQLQQFRAD PSTASLLLCP GGLDHKLNLR GKAWG
Greater than 80% as determined by SDS-PAGE.
9220
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers