missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Testis-expressed sequence 12 Protein
A DNA sequence encoding the Homo sapiens (Human) Testis-expressed sequence 12 protein, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_112565.1 |
---|---|
Gene ID (Entrez) | 56158 |
Name | Testis-expressed sequence 12 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055730
|
enQuireBio™
QP6773-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075730
|
enQuireBio™
QP6773-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065730
|
enQuireBio™
QP6773-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085730
|
enQuireBio™
QP6773-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095730
|
enQuireBio™
QP6773-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005740
|
enQuireBio™
QP6773-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_112565.1 | |
Testis-expressed sequence 12 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
56158 | |
Research Use Only | |
TEX12 | |
Recombinant Protein | |
E. coli | |
MMANHLVKPD NRNCKRPREL ESPVPDSPQL SSLGKSDSSF SEISGLFYKD EALEKDLNDV SKEINLMLST YAKLLSERAA VDASYIDEID ELFKEANAIE NFLIQKREFL RQRFTVIANT LHR | |
Greater than 90% as determined by SDS-PAGE. |